4-Methyl-N-[3-(trifluoromethyl)phenyl]benzamide 97%

Warning

H57103-MD H57103-MD H57103-03 H57103-03
AAH57103-03EA 194.25 USD
CAAAH57103-03 AAH57103-03 CAAAH57103-MD AAH57103-MD
4-Methyl-N-[3-(trifluoromethyl)phenyl]benzamide 97%
4-Methyl-N-[3-(trifluoromethyl)phenyl]benzamide
Formula: C₁₅H₁₂F₃NO
Storage Temperature: Ambient
MDL Number: MFCD00423044
CAS Number: 144918-48-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR