3-Fluoro-N-(4-fluorobenzyl)aniline 97%

Warning

H56989-03 H56989-MD H56989-03 H56989-MD
AAH56989-03EA 420.69 USD
AAH56989-03 AAH56989-MD CAAAH56989-MD CAAAH56989-03
3-Fluoro-N-(4-fluorobenzyl)aniline 97%
3-Fluoro-N-(4-fluorobenzyl)aniline
Formula: C₁₃H₁₁F₂N
MW: 219.23
Storage Temperature: Ambient
MDL Number: MFCD11144475
CAS Number: 1019613-18-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR