Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

4-Chloro-N-(4-fluorobenzyl)aniline 97%

Supplier: Alfa Aesar
Warning

H56981-MD H56981-03 H56981-03 H56981-MD
AAH56981-MDEA 54.68 USD InStock
AAH56981-MD CAAAH56981-03 AAH56981-03 CAAAH56981-MD
4-Chloro-N-(4-fluorobenzyl)aniline 97%
4-Chloro-N-(4-fluorobenzyl)aniline


Formula: C₁₃H₁₁ClFN
MW: 235,69
Storage Temperature: Ambient
MDL Number: MFCD03210761
CAS Number: 356531-46-9
EINECS: 000-000-0
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty