Dimethyl-trans-cyclohexane-1,4-dicarboxylate 99%
|
Warning
|
Synonyms:
Dimethyl trans-1,4-Cyclohexanedicarboxylate
, Dimethyl trans-1,4-cyclohexanedicarboxylate
H56938-03
H56938-06
H56938-06
H56938-03
AAH56938-03EA
38.61
USD
CAAAH56938-06
AAH56938-03
CAAAH56938-03
AAH56938-06
Dimethyl-trans-cyclohexane-1,4-dicarboxylate 99%
Dimethyl trans-cyclohexane-1,4-dicarboxylate
Formula:
C₁₀H₁₆O₄ MW: 200.23 g/mol Melting Pt: 67…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063917 CAS Number: 3399-22-2 |
Specification Test Results
Assay Minimum | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...