2-Chloro-N-methylbenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H56910-03 H56910-MD H56910-03 H56910-MD
AAH56910-03EA 213.99 USD
AAH56910-03 AAH56910-MD CAAAH56910-03 CAAAH56910-MD
2-Chloro-N-methylbenzamide 97%
2-Chloro-N-methylbenzamide
Formula: C₈H₈ClNO
MW: 169.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00032667
CAS Number: 3400-31-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR