trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile 99%

Warning

Synonyms: 4'-(trans-4-Pentylcyclohexyl)-[1,1'-biphenyl]-4-carbonitrile

H56850-03 H56850-06 H56850-06 H56850-03
AAH56850-03EA 48.61 USD
CAAAH56850-06 CAAAH56850-03 AAH56850-03 AAH56850-06
trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile 99%
trans-4-[4-(4-Pentylcyclohexyl)phenyl]benzonitrile
Formula: C₂₄H₂₉N
MW: 331.50 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD01076317
CAS Number: 68065-81-6
EINECS: 268-333-6

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR