Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxy-N-methylbenzamide ≥97%

Supplier: Alfa Aesar
Warning

H56836-03 H56836-MD H56836-MD H56836-03
AAH56836-03EA 451.14 USD InStock
AAH56836-03 CAAAH56836-MD AAH56836-MD CAAAH56836-03
2-Methoxy-N-methylbenzamide ≥97%
2-Methoxy-N-methylbenzamide


Formula: C₉H₁₁NO₂
MW: 165.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01578285
CAS Number: 3400-35-9
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty