4-Methoxy-N-(1-naphthyl)benzenesulfonamide 97%

Warning

H56822-MD H56822-03 H56822-MD H56822-03
AAH56822-MDEA 145.41 USD
CAAAH56822-03 CAAAH56822-MD AAH56822-MD AAH56822-03
4-Methoxy-N-(1-naphthyl)benzenesulfonamide 97%
4-Methoxy-N-(1-naphthyl)benzenesulfonamide
Formula: C₁₇H₁₅NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00682767
CAS Number: 56799-97-4

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR