N-(2-Fluorobenzyl)aniline 97%

Warning

H56725-03 H56725-03 H56725-MD H56725-MD
AAH56725-03EA 231.99 USD
AAH56725-03 AAH56725-MD CAAAH56725-MD CAAAH56725-03
N-(2-Fluorobenzyl)aniline 97%
N-(2-Fluorobenzyl)aniline
Formula: C₁₃H₁₂FN
MW: 201.24
Storage Temperature: Ambient
MDL Number: MFCD06408526
CAS Number: 940362-32-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR