N-(4-Fluorobenzyl)-3-methoxyaniline 97%

Warning

H56708-03 H56708-MD H56708-03 H56708-MD
AAH56708-MDEA 104.67 USD
AAH56708-MD CAAAH56708-MD AAH56708-03 CAAAH56708-03
N-(4-Fluorobenzyl)-3-methoxyaniline 97%
N-(4-Fluorobenzyl)-3-methoxyaniline
Formula: C₁₄H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD03210777
CAS Number: 356531-63-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR