4-Bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzenesulfonamide 97%
|
Warning
|
H56605-03
H56605-MD
H56605-03
H56605-MD
AAH56605-03EA
416.25
USD
CAAAH56605-MD
AAH56605-03
AAH56605-MD
CAAAH56605-03
4-Bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzenesulfonamide 97%
4-Bromo-N-(4-chloro-2,5-dimethoxyphenyl)benzenesulphonamide
Formula:
C₁₄H₁₃BrClNO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00588020 CAS Number: 346721-67-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...