4-Ethoxyphenyl-trans-4-(4-n-pentylcyclohexyl)benzoate 99%

H56493-06 H56493-06 H56493-03 H56493-03
AAH56493-06EA 289.71 USD
CAAAH56493-06 AAH56493-06 AAH56493-03 CAAAH56493-03
4-Ethoxyphenyl-trans-4-(4-n-pentylcyclohexyl)benzoate 99%
4-Ethoxyphenyl-trans-4-(4-n-pentylcyclohexyl)benzoate
Formula: C₂₆H₃₄O₃
MW: 394.555 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09952999
CAS Number: 95973-51-6

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR