N-Methyl-4-methylbenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H56446-03 H56446-MD H56446-03 H56446-MD
AAH56446-MDEA 96.49 USD
AAH56446-MD AAH56446-03 CAAAH56446-MD CAAAH56446-03
N-Methyl-4-methylbenzamide 97%
N-Methyl-4-methylbenzamide
Formula: C₉H₁₁NO
MW: 149.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043627
CAS Number: 18370-11-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR