4-Ethylphenyl-trans-4-(4-n-pentylcyclohexyl)benzoate 99%

H56418-03 H56418-06 H56418-06 H56418-03
AAH56418-03EA 58.48 USD
CAAAH56418-06 AAH56418-03 CAAAH56418-03 AAH56418-06
4-Ethylphenyl-trans-4-(4-n-pentylcyclohexyl)benzoate 99%
4-Ethylphenyl-trans-4-(4-n-pentylcyclohexyl)benzoate
Formula: C₂₆H₃₄O₂
MW: 378.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16879074
CAS Number: 91223-44-8

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR