N-Benzyl-4-fluorobenzylamine 97%

Warning

H56362-03 H56362-MD H56362-03 H56362-MD
AAH56362-MDEA 155.98 USD
CAAAH56362-MD AAH56362-MD AAH56362-03 CAAAH56362-03
N-Benzyl-4-fluorobenzylamine 97%
N-Benzyl-4-fluorobenzylamine
Formula: C₁₄H₁₄FN
MW: 215.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00716882
CAS Number: 55096-88-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR