N-Methyl-3-methylbenzamide 97%

Warning

H56299-MD H56299-03 H56299-MD H56299-03
AAH56299-03EA 462.87 USD
AAH56299-03 CAAAH56299-03 CAAAH56299-MD AAH56299-MD
N-Methyl-3-methylbenzamide 97%
N-Methyl-3-methylbenzamide
Formula: C₉H₁₁NO
MW: 149.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01211937
CAS Number: 74786-81-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR