4-Chloro-N-(4-fluorobenzyl)-2-methylaniline 97%

Supplier: Thermo Scientific Chemicals

Warning

H56258-03 H56258-MD H56258-MD H56258-03
AAH56258-MDEA 131 USD
AAH56258-MD CAAAH56258-03 AAH56258-03 CAAAH56258-MD
4-Chloro-N-(4-fluorobenzyl)-2-methylaniline 97%
4-Chloro-N-(4-fluorobenzyl)-2-methylaniline
Formula: C₁₄H₁₃ClFN
Storage Temperature: Ambient
MDL Number: MFCD11119360
CAS Number: 1036628-11-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR