4-Methoxy-N-(4-methylbenzyl)benzenesulfonamide 97%
|
Warning
|
Synonyms:
4-Methoxy-N-(4-methyl-benzyl)-benzenesulfonamide
H56245-03
H56245-03
H56245-MD
H56245-MD
AAH56245-MDEA
102.56
USD
AAH56245-MD
CAAAH56245-03
AAH56245-03
CAAAH56245-MD
4-Methoxy-N-(4-methylbenzyl)benzenesulfonamide 97%
4-Methoxy-N-(4-methylbenzyl)benzenesulfonamide
Formula:
C₁₅H₁₇NO₃S MW: 291.36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01213791 CAS Number: 346696-69-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...