N-(4-Fluorobenzyl)-4-methoxyaniline 97%
|
Warning
|
H56102-MD
H56102-03
H56102-MD
H56102-03
AAH56102-MDEA
145.41
USD
AAH56102-MD
CAAAH56102-MD
AAH56102-03
CAAAH56102-03
N-(4-Fluorobenzyl)-4-methoxyaniline 97%
N-(4-Fluorobenzyl)-4-methoxyaniline
Formula:
C₁₄H₁₄FNO Storage Temperature: Ambient |
MDL Number:
MFCD03210757 CAS Number: 356531-43-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...