3-Ethynylthiophene 96%

Supplier: Thermo Scientific Chemicals

Danger

H55898-MD H55898-06 H55898-03 H55898-06 H55898-MD H55898-03
AAH55898-03EA 60.39 USD
CAAAH55898-03 CAAAH55898-MD CAAAH55898-06 AAH55898-03 AAH55898-MD AAH55898-06
3-Ethynylthiophene 96%
3-Ethynylthiophene
Formula: C₆H₄S
MW: 108.16 g/mol
Boiling Pt: 152…153 °C
Density: 1.098
Flash Pt: 46 °C(115 °F)
Storage Temperature: Ambient
MDL Number: MFCD04039973
CAS Number: 67237-53-0
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR