4-Cyano-4'-hexylbiphenyl 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Cyano-4'-n-hexylbiphenyl
, 4-Hexyl-[1,1-biphenyl]-4-carbonitrile
H55825-03
H55825-MD
H55825-06
H55825-06
H55825-03
H55825-MD
AAH55825-MDEA
30.97
USD
CAAAH55825-03
AAH55825-MD
AAH55825-06
AAH55825-03
CAAAH55825-MD
CAAAH55825-06
4-Cyano-4'-hexylbiphenyl 97%
4-Cyano-4'-hexylbiphenyl
Formula:
C₁₉H₂₁N MW: 263.38 g/mol Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075144 CAS Number: 41122-70-7 EINECS: 255-228-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...