Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Statsafe™ 6000

Supplier: Alfa Aesar
Danger

H55787-22 H55787-36 H55787-14 H55787-22 H55787-36 H55787-14
AAH55787-14EA 54.43 USD InStock
CAAAH55787-22 CAAAH55787-36 AAH55787-14 AAH55787-22 AAH55787-36 CAAAH55787-14
Statsafe™ 6000
Statsafe™ 6000
Applications: Static dissipator used in many chemical processing operations to reduce the risk of static build up and consequent damage and loss.
Suitable additive to increase hydrocarbon conductivity and to reduce electrostatic hazards in a variety of applications.


Boiling Pt: 94°C
Density: 0,805
Flash Pt: <-5°C (-23°F)
Storage Temperature: Ambient
UN: 2924
ADR: 3,II
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty