2',4'-Dimethylacetanilide 98%

Warning

Synonyms: N-(2,4-Dimethylphenyl)acetamide

H55605-06 H55605-14
AAH55605-06EA 38.94 USD
AAH55605-06 AAH55605-14
2',4'-Dimethylacetanilide 98%
2',4'-Dimethylacetanilide
Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00026125
CAS Number: 2050-43-3
EINECS: 218-088-6

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR