L-Threonyl diphenylborinate 95%

Synonyms: Diphenylborinic acid L-threonyl ester(2S,3R)-2-Amino-3-hydroxybutyryl diphenylborinate

H55599-03 H55599-MD H55599-03 H55599-MD
AAH55599-MDEA 65.07 USD
CAAAH55599-03 CAAAH55599-MD AAH55599-MD AAH55599-03
L-Threonyl diphenylborinate 95%
L-Threonyl diphenylborinate
Formula: C₁₆H₁₈BNO₃
MW: 283.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22125157
CAS Number: 16655-44-0

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR