1-(3-Methoxyphenyl)piperazine 95%

Warning

H55450-03 H55450-06 H55450-14 H55450-03 H55450-06 H55450-14
AAH55450-06EA 75.63 USD
AAH55450-06 AAH55450-03 AAH55450-14 CAAAH55450-03 CAAAH55450-14 CAAAH55450-06
1-(3-Methoxyphenyl)piperazine 95%
1-(3-Methoxyphenyl)piperazine
Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
Boiling Pt: 150 °C/0.5 mm
Density: 1.114
Flash Pt: 91 °C(196 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040733
CAS Number: 16015-71-7
EINECS: 240-154-8
ADR: 8,III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR