L-Serinyl diphenylborinate 95%

Synonyms: Diphenylborinic acid L-serinyl ester(S)-2-Amino-3-hydroxypropionyl diphenylborinate

H55447-03 H55447-MD H55447-03 H55447-MD
AAH55447-03EA 493.75 USD
AAH55447-03 CAAAH55447-MD CAAAH55447-03 AAH55447-MD
L-Serinyl diphenylborinate 95%
L-Serinyl diphenylborinate
Formula: C₁₅H₁₆BNO₃
MW: 269.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22125144
CAS Number: 16657-63-9

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR