N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide 97%
|
Warning
|
H55330-03
H55330-MD
H55330-03
H55330-MD
AAH55330-03EA
330.78
USD
AAH55330-03
AAH55330-MD
CAAAH55330-03
CAAAH55330-MD
N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide 97%
N-(3,4-Dimethylphenyl)-4-fluorobenzenesulphonamide
Formula:
C₁₄H₁₄FNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD00784407 CAS Number: 288154-65-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...