4-Nitrobenzenediazonium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Danger

H55315-03 H55315-03 H55315-14 H55315-06 H55315-14 H55315-06
AAH55315-03EA 26.52 USD
AAH55315-03 AAH55315-14 CAAAH55315-06 CAAAH55315-03 CAAAH55315-14 AAH55315-06
4-Nitrobenzenediazonium tetrafluoroborate 97%
4-Nitrobenzenediazonium tetrafluoroborate
Formula: C₆H₄BF₄N₃O₂
MW: 236.92 g/mol
Melting Pt: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00012005
CAS Number: 456-27-9
EINECS: 207-261-1
UN: 2923
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR