1-(2-Methoxyethyl)piperazine 97%

Danger

H55278-03 H55278-03 H55278-MD H55278-06 H55278-MD H55278-06
AAH55278-03EA 65.52 USD
CAAAH55278-06 CAAAH55278-MD AAH55278-03 AAH55278-06 AAH55278-MD CAAAH55278-03
1-(2-Methoxyethyl)piperazine 97%
1-(2-Methoxyethyl)piperazine
Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
Boiling Pt: 193…194 °C
Density: 0.970
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191214
CAS Number: 13484-40-7
UN: 2735
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR