N-(Trimethylsilyl)morpholine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Trimethylsilyl)morpholine

H55229-03 H55229-06 H55229-14 H55229-03 H55229-06 H55229-14
AAH55229-03EA 18.65 USD
AAH55229-03 CAAAH55229-14 CAAAH55229-03 AAH55229-14 CAAAH55229-06 AAH55229-06
N-(Trimethylsilyl)morpholine 97%
N-(Trimethylsilyl)morpholine
Formula: C₇H₁₇NOSi
MW: 159.30 g/mol
Boiling Pt: 160 °C
Density: 0.897
Flash Pt: 29 °C(84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010098
CAS Number: 13368-42-8
UN: 2924
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR