L-Isoleucyl diphenylborinate 95%

Synonyms: Diphenylborinic acid L-isoleucyl ester

H55184-03 H55184-MD H55184-MD H55184-03
AAH55184-03EA 509.49 USD
CAAAH55184-03 AAH55184-03 AAH55184-MD CAAAH55184-MD
L-Isoleucyl diphenylborinate 95%
L-Isoleucyl diphenylborinate
Formula: C₁₈H₂₂BNO₂
MW: 295.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22125121
CAS Number: 124383-92-2

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR