N-tert-Butyl-4-methylbenzenesulfonamide 97%
|
Warning
|
Synonyms:
N-tert-Butyl-p-toluenesulfonamide
H55126-MB
H55126-MB
AAH55126-MBEA
362.97
USD
AAH55126-MB
CAAAH55126-MB
N-tert-Butyl-4-methylbenzenesulfonamide 97%
N-tert-Butyl-4-methylbenzenesulfonamide
Formula:
C₁₁H₁₇NO₂S MW: 227.32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00026270 CAS Number: 2849-81-2 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...