2,4,6-Trimethylphenylmagnesium bromide 1 M in 2-MeTHF (2-Methyltetrahydrofuran)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Mesitylmagnesium bromide
H54282-AD
H54282-AE
H54282-AE
AAH54282-AEEA
388.99
USD
CAAAH54282-AE
AAH54282-AE
AAH54282-AD
2,4,6-Trimethylphenylmagnesium bromide 1 M in 2-MeTHF (2-Methyltetrahydrofuran)
2,4,6-Trimethylphenylmagnesium bromide
Formula:
C₉H₁₁BrMg MW: 223.396 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009663 CAS Number: 2633-66-1 UN: 2924 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...