4-Methoxybenzylmagnesium chloride 0.25 M in 2-MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H54078-AE H54078-AE
AAH54078-AEEA 438.99 USD
CAAAH54078-AE AAH54078-AE
4-Methoxybenzylmagnesium chloride 0.25 M in 2-MeTHF (2-methyltetrahydrofuran)
4-Methoxybenzylmagnesium chloride
Formula: C₈H₉ClMgO
MW: 180.914 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311491
CAS Number: 38769-92-5
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR