3-Chloro-4-methylphenylmagnesium bromide 0.50 M in 2-MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H54075-AE H54075-AE
AAH54075-AEEA 231.99 USD
CAAAH54075-AE AAH54075-AE
3-Chloro-4-methylphenylmagnesium bromide 0.50 M in 2-MeTHF (2-methyltetrahydrofuran)
3-Chloro-4-methylphenylmagnesium bromide
Formula: C₇H₆BrClMg
MW: 229.784 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11553191
CAS Number: 515158-85-7
UN: 2924
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR