1,8-Nonadiene 99%

Warning

Synonyms: Nona-1,8-diene

H53502-03 H53502-06 H53502-14 H53502-03 H53502-14 H53502-06
AAH53502-03EA 26.39 USD
AAH53502-03 AAH53502-14 CAAAH53502-06 CAAAH53502-03 CAAAH53502-14 AAH53502-06
1,8-Nonadiene 99%
1,8-Nonadiene
Formula: C₉H₁₆
MW: 124.23 g/mol
Boiling Pt: 141…142 °C
Density: 0.74
Flash Pt: 26 °C(79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008669
CAS Number: 4900-30-5
EINECS: 225-527-5
UN: 3295
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR