3-(Trimethylsilyl)propiolaldehyde diethyl acetal 97%

Supplier: Thermo Scientific Chemicals
Warning

H53397-06 H53397-14 H53397-14 H53397-06
AAH53397-14EA 238.07 USD
AAH53397-14 CAAAH53397-06 CAAAH53397-14 AAH53397-06
3-(Trimethylsilyl)propiolaldehyde diethyl acetal 97%
3-(Trimethylsilyl)propiolaldehyde diethyl acetal
Formula: C₁₀H₂₀O₂Si
MW: 200.353 g/mol
Boiling Pt: 170…172 °C
Flash Pt: 40 °C(104 °F)
Storage Temperature: Ambient
MDL Number: MFCD08275372
CAS Number: 87219-80-5
UN: 1993
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR