Piperazine hexahydrate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Piperazine hydrate
H37069-0I
H37069-0B
H37069-30
AAH37069-30EA
54
USD
AAH37069-30
AAH37069-0I
AAH37069-0B
Piperazine hexahydrate 97+%
Piperazine hexahydrate
Formula:
C₄H₁₀N₂·6H₂O MW: 194.23 g/mol Boiling Pt: 125…130 °C Melting Pt: 42…46 °C Flash Pt: >100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00149389 CAS Number: 142-63-2 EINECS: 203-808-3 UN: 2579 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...