Ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate hydrate 95%

Warning

H35813-03 H35813-03 H35813-MD H35813-MD
AAH35813-MDEA 101.56 USD
AAH35813-MD AAH35813-03 CAAAH35813-03 CAAAH35813-MD
Ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate hydrate 95%
Ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate hydrate
Formula: C₁₀H₁₁N₃O₂·H₂O
MW: 205.22 g/mol (anhy)
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD22989346
CAS Number: 1000017-97-9

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR