6-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid 95%
|
Warning
|
H35790-MD
H35790-MD
H35790-03
H35790-03
AAH35790-MDEA
52.84
USD
AAH35790-MD
CAAAH35790-MD
CAAAH35790-03
AAH35790-03
6-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid 95%
6-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Formula:
C8H5BrN2O2 MW: 241.05 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD22989345 CAS Number: 749849-14-7 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...