7-Bromoimidazo[1,2-a]pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H35721-03
H35721-MD
H35721-MD
H35721-03
AAH35721-MDEA
130.17
USD
CAAAH35721-03
AAH35721-MD
AAH35721-03
CAAAH35721-MD
7-Bromoimidazo[1,2-a]pyridine 95%
7-Bromoimidazo[1,2-a]pyridine
Formula:
C₇H₅BrN₂ MW: 197.03 g/mol Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD07778353 CAS Number: 808744-34-5 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...