Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
|
Warning
|
Synonyms:
Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
H35652-MD
H35652-MD
H35652-03
H35652-03
AAH35652-03EA
227.55
USD
CAAAH35652-03
AAH35652-03
CAAAH35652-MD
AAH35652-MD
Ethyl-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Formula:
C₁₁H₉F₃N₂O₂ MW: 258.20 g/mol Melting Pt: 139…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD11100212 CAS Number: 860457-99-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...