Methyl-3-chloroimidazo[1,2-a]pyridine-6-carboxylate 95%
|
Warning
|
H35637-03
H35637-MD
H35637-MD
H35637-03
AAH35637-03EA
254.19
USD
CAAAH35637-03
CAAAH35637-MD
AAH35637-03
AAH35637-MD
Methyl-3-chloroimidazo[1,2-a]pyridine-6-carboxylate 95%
Methyl 3-chloroimidazo[1,2-a]pyridine-6-carboxylate
Formula:
C₉H₇ClN₂O₂ MW: 210.62 Melting Pt: 119…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD07021500 CAS Number: 900019-34-3 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...