Ethyl-3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%
|
Warning
|
H35508-MD
H35508-03
H35508-MD
H35508-03
AAH35508-03EA
126.54
USD
CAAAH35508-MD
AAH35508-03
AAH35508-MD
CAAAH35508-03
Ethyl-3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formula:
C₁₀H₈BrClN₂O₂ MW: 303.54 Melting Pt: 139…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD06496148 CAS Number: 861208-16-4 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...