3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid 95%

Warning

H35479-03 H35479-MD H35479-MD H35479-03
AAH35479-MDEA 100.09 USD
AAH35479-MD CAAAH35479-MD CAAAH35479-03 AAH35479-03
3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid 95%
3-bromoimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Formula: C₈H₅BrN₂O₂
MW: 241.044 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD22989342
CAS Number: 354548-73-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR