Ethyl-3-chloroimidazo[1,2-a]pyridine-6-carboxylate 95%
|
Warning
|
H35396-03
H35396-03
H35396-MD
H35396-MD
AAH35396-03EA
310.8
USD
CAAAH35396-MD
CAAAH35396-03
AAH35396-03
AAH35396-MD
Ethyl-3-chloroimidazo[1,2-a]pyridine-6-carboxylate 95%
Ethyl 3-chloroimidazo[1,2-a]pyridine-6-carboxylate
Formula:
C₁₀H₉ClN₂O₂ MW: 224.65 Melting Pt: 101…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD21100199 CAS Number: 1427460-48-7 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...