Ethyl imidazo[1,2-a]pyridine-2-carboxylate 95%
|
Warning
|
H35384-03
H35384-09
H35384-09
H35384-03
AAH35384-09EA
394.05
USD
AAH35384-09
CAAAH35384-09
AAH35384-03
CAAAH35384-03
Ethyl imidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl imidazo[1,2-a]pyridine-2-carboxylate
Formula:
C₁₀H₁₀N₂O₂ Melting Pt: 78…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD02325394 CAS Number: 38922-77-9 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...