Methyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate 95%
|
Warning
|
H35331-MD
H35331-03
H35331-03
H35331-MD
AAH35331-03EA
420.69
USD
AAH35331-03
CAAAH35331-MD
AAH35331-MD
CAAAH35331-03
Methyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate 95%
Methyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate
Formula:
C₉H₇ClN₂O₂ MW: 210.62 Melting Pt: 151…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD07021508 CAS Number: 900019-38-7 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...