8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate 95%

Warning

Synonyms: 8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate

H35323-03 H35323-MD H35323-03 H35323-MD
AAH35323-MDEA 157.62 USD
AAH35323-MD CAAAH35323-03 AAH35323-03 CAAAH35323-MD
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate 95%
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylic acid hydrate
Formula: C₈H₄BrClN₂O₂·xH₂O
MW: 275.49 g/mol (anhy)
Melting Pt: 215…217 °C
Storage Temperature: Ambient
MDL Number: MFCD22682816
CAS Number: 1858255-62-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR