3-Iodopyrazolo[1,5-a]pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H35255-03 H35255-03
AAH35255-03EA 548.99 USD
CAAAH35255-03 AAH35255-03
3-Iodopyrazolo[1,5-a]pyridine 97%
3-Iodopyrazolo[1,5-a]pyridine
Formula: C₇H₅IN₂
MW: 244.035 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD20483547
CAS Number: 19363-99-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR